Assay Details
Gene Symbol: Erich2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024311.1 | NP_001019482.1 | 6 - 7 | 923 | 73 | |
XM_006234340.2 | XP_006234402.1 | 6 - 7 | 2029 | 73 | ||
GenBank mRNA | BC082109.1 | - | 6 - 7 | 923 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
