Assay Details
Gene Symbol: Gpat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168529.1 | NP_001162001.1 | 19 - 20 | 2294 | 95 | |
XM_006234956.2 | XP_006235018.1 | 22 - 23 | 2692 | 95 | ||
XM_008762158.1 | XP_008760380.1 | 19 - 20 | 3036 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
