Assay Details
Gene Symbol: Idh3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053581.1 | NP_446033.1 | 10 - 11 | 1020 | 66 | |
XM_006235048.2 | XP_006235110.1 | 10 - 11 | 1057 | 66 | ||
GenBank mRNA | AB047540.1 | - | 3 - 4 | 289 | 66 | |
BC079113.1 | - | 10 - 11 | 1020 | 7126029 | 66 | |
FQ215180.1 | - | 10 - 11 | 1024 | 66 | ||
FQ215184.1 | - | 10 - 11 | 1011 | 66 | ||
FQ215350.1 | - | 10 - 11 | 1020 | 66 | ||
FQ217667.1 | - | 4 - 5 | 378 | 66 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
