Assay Details
Gene Symbol: Idh3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053581.1 | NP_446033.1 | 3 - 4 | 229 | 70 | |
XM_006235048.2 | XP_006235110.1 | 3 - 4 | 266 | 70 | ||
GenBank mRNA | BC079113.1 | - | 3 - 4 | 229 | 70 | |
FQ214820.1 | - | 3 - 4 | 227 | 70 | ||
FQ215180.1 | - | 3 - 4 | 233 | 70 | ||
FQ215184.1 | - | 3 - 4 | 221 | 70 | ||
FQ215350.1 | - | 3 - 4 | 229 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
