Assay Details
Gene Symbol: Sirpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234951.2 | XP_006235013.1 | 1 - 2 | 615 | 64 | |
XM_006234952.2 | XP_006235014.1 | 2 - 3 | 977 | 64 | ||
XM_006234953.2 | XP_006235015.1 | 2 - 3 | 1159 | 64 | ||
GenBank mRNA | AF055065.1 | - | 2 - 3 | 181 | 64 | |
BC099773.1 | - | 2 - 3 | 253 | 64 | ||
D38468.1 | - | 1 - 2 | 372 | 64 | ||
D85183.1 | - | 1 - 2 | 129 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
