Assay Details
Gene Symbol: Cry2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133405.2 | NP_596896.2 | 3 - 4 | 461 | 75 | |
GenBank mRNA | AY033877.1 | - | 3 - 4 | 471 | 75 | |
BC166416.1 | - | 3 - 4 | 461 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
