Assay Details
Gene Symbol: Slc39a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039196.1 | NP_001034285.1 | 3 - 4 | 585 | 59 | |
XM_006234501.2 | XP_006234563.1 | 3 - 4 | 791 | 59 | ||
XM_006234502.2 | XP_006234564.1 | 3 - 4 | 795 | 59 | ||
GenBank mRNA | BC112321.1 | - | 3 - 4 | 585 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
