Assay Details
Gene Symbol: Ttbk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107766.1 | NP_001101236.1 | 1 - 2 | 116 | 88 | |
XM_006234835.2 | XP_006234897.1 | 1 - 2 | 382 | 88 | ||
XM_006234836.2 | XP_006234898.1 | 1 - 2 | 382 | 88 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001107766.1, XM_006234835.2, XM_006234836.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
