Assay Details
Gene Symbol: Ttbk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107766.1 | NP_001101236.1 | 3 - 4 | 397 | 76 | |
XM_006234833.2 | XP_006234895.1 | 4 - 5 | 737 | 76 | ||
XM_006234834.2 | XP_006234896.1 | 4 - 5 | 380 | 76 | ||
XM_006234835.2 | XP_006234897.1 | 3 - 4 | 663 | 76 | ||
XM_006234837.2 | XP_006234899.1 | 4 - 5 | 383 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
