Assay Details
Gene Symbol: Srp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106497.1 | NP_001099967.1 | 3 - 4 | 291 | 94 | |
GenBank mRNA | BC166931.1 | - | 3 - 4 | 247 | 94 | |
FQ210704.1 | - | 2 - 3 | 191 | 94 | ||
FQ210780.1 | - | 2 - 3 | 138 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
