Assay Details
Gene Symbol: Tcp11l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109202.1 | NP_001102672.1 | 1 - 2 | 193 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109202.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
