Assay Details
Gene Symbol: Tcp11l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109202.1 | NP_001102672.1 | 8 - 9 | 1366 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
