Assay Details
Gene Symbol: Hao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107780.2 | NP_001101250.1 | 5 - 6 | 828 | 92 | |
GenBank mRNA | BC158804.1 | - | 5 - 6 | 828 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
