Assay Details
Gene Symbol: Hao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107780.2 | NP_001101250.1 | 7 - 8 | 1053 | 99 | |
GenBank mRNA | BC158804.1 | - | 7 - 8 | 1053 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P340172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377561 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172001 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208062 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10033 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158510 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P340172 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P340172 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P340172 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P340172 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11477 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10850194 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850195 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850191 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850192 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850197 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850193 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850196 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850198 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10850190 | - | - |
Rat Genome 230 2.0 Array | 1394112_PM_at | - | - |
Rat Genome 230 2.0 Array | 1394112_at | - | - |
Affymetrix HT Rat Focus Array | 1394112_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1394112_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300032995.rn.1 | TC0300004239.rn.1 | RTA TC |
Rat Expression Set 230 | 1394112_at | - | - |
Rat Expression Set 230 | 1394112_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10850190 | - | - |
Rat Genome U34 Set | rc_AA945123_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_2038974 | - | - |
