Assay Details
Gene Symbol: Slc1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035233.1 | NP_001030310.1 | 836 | 71 | ||
NM_017215.2 | NP_058911.2 | 836 | 71 | |||
GenBank mRNA | AF297648.1 | - | 748 | 71 | ||
AF451299.1 | - | 208 | 71 | |||
AF465909.1 | - | 729 | 71 | |||
AY044832.1 | - | 725 | 71 | |||
JN132400.1 | - | 911 | 71 | |||
U15098.1 | - | 785 | 71 | |||
X67857.1 | - | 823 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
