Assay Details
Gene Symbol: Slc1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035233.1 | NP_001030310.1 | 1196 | 134 | ||
NM_017215.2 | NP_058911.2 | 1196 | 134 | |||
GenBank mRNA | AF297648.1 | - | 1108 | 134 | ||
AF451299.1 | - | 568 | 134 | |||
AF465909.1 | - | 1089 | 134 | |||
AY044832.1 | - | 1085 | 134 | |||
AY069978.1 | - | 387 | 134 | |||
JN132400.1 | - | 1271 | 134 | |||
U15098.1 | - | 1145 | 134 | |||
X67857.1 | - | 1183 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
