Assay Details
Gene Symbol: Ddb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271346.1 | NP_001258275.1 | 1 - 2 | 294 | 61 | |
XM_006234499.2 | XP_006234561.1 | 1 - 2 | 327 | 61 | ||
GenBank mRNA | FQ228020.1 | - | 1 - 2 | 294 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
MicroArray Probes:
