Assay Details
Gene Symbol: Ddb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271346.1 | NP_001258275.1 | 5 - 6 | 870 | 77 | |
XM_006234498.2 | XP_006234560.1 | 5 - 6 | 863 | 77 | ||
XM_006234499.2 | XP_006234561.1 | 5 - 6 | 903 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
