Assay Details
Gene Symbol: Rmdn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014046.2 | NP_001014068.1 | 1 - 2 | 81 | 132 | |
GenBank mRNA | DN931790.1 | - | 1 - 2 | 113 | 132 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001014046.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
