Assay Details
Gene Symbol: Blvra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234894.2 | XP_006234956.1 | 2 - 3 | 248 | 70 | |
XM_006234895.2 | XP_006234957.1 | 2 - 3 | 233 | 70 | ||
GenBank mRNA | BC078766.1 | - | 1 - 2 | 120 | 7126157 | 70 |
FQ213399.1 | - | 2 - 3 | 187 | 70 | ||
FQ230935.1 | - | 1 - 2 | 67 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC078766.1, XM_006234894.2, XM_006234895.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
