Assay Details
Gene Symbol: Mmp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053963.2 | NP_446415.2 | 2 - 3 | 377 | 77 | |
GenBank mRNA | BC088120.1 | - | 2 - 3 | 377 | 77 | |
X98517.1 | - | 2 - 3 | 377 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
