Assay Details
Gene Symbol: Eif3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013095.1 | NP_001013113.1 | 6 - 7 | 416 | 86 | |
GenBank mRNA | BC086383.1 | - | 6 - 7 | 416 | 86 | |
FQ213005.1 | - | 6 - 7 | 457 | 86 | ||
FQ225883.1 | - | 6 - 7 | 449 | 86 | ||
FQ226276.1 | - | 6 - 7 | 448 | 86 | ||
FQ229703.1 | - | 6 - 7 | 447 | 86 | ||
FQ230888.1 | - | 6 - 7 | 442 | 86 | ||
FQ232595.1 | - | 6 - 7 | 446 | 86 | ||
FQ232661.1 | - | 6 - 7 | 446 | 86 | ||
FQ234018.1 | - | 6 - 7 | 449 | 86 | ||
FQ234727.1 | - | 4 - 5 | 298 | 86 | ||
FQ234921.1 | - | 6 - 7 | 447 | 86 | ||
FQ234929.1 | - | 6 - 7 | 449 | 86 | ||
FQ235264.1 | - | 3 - 4 | 230 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
