Assay Details
Gene Symbol: Mrpl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108754.1 | NP_001102224.1 | 1 - 2 | 255 | 59 | |
GenBank mRNA | FQ215152.1 | - | 1 - 2 | 121 | 59 | |
FQ226445.1 | - | 1 - 2 | 121 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
