Assay Details
Gene Symbol: Akr1e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008342.1 | NP_001008343.1 | 7 - 8 | 762 | 94 | |
XM_008771865.1 | XP_008770087.1 | 6 - 7 | 665 | 94 | ||
GenBank mRNA | BC086397.1 | - | 7 - 8 | 762 | 7302893 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
