Assay Details
Gene Symbol: Il15ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728509.4 | XP_002728555.2 | 1 - 2 | 681 | 85 | |
XM_006254225.2 | XP_006254287.1 | 1 - 2 | 676 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
