Assay Details
Gene Symbol: Il15ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728509.4 | XP_002728555.2 | 2 - 3 | 868 | 67 | |
XM_006254225.2 | XP_006254287.1 | 2 - 3 | 863 | 67 | ||
GenBank mRNA | DQ157696.1 | - | 2 - 3 | 155 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
