Assay Details
Gene Symbol: Il15ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728509.4 | XP_002728555.2 | 3 - 4 | 969 | 83 | |
GenBank mRNA | DQ157696.1 | - | 3 - 4 | 256 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
