Assay Details
Gene Symbol: Il15ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728509.4 | XP_002728555.2 | 6 - 7 | 1276 | 72 | |
XM_006254225.2 | XP_006254287.1 | 5 - 6 | 1073 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
