Assay Details
Gene Symbol: Sec61a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170343.1 | NP_001163814.1 | 2 - 3 | 149 | 111 | |
XM_006254257.2 | XP_006254319.1 | 2 - 3 | 249 | 111 | ||
GenBank mRNA | CK470805.1 | - | 2 - 3 | 172 | 111 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001170343.1, XM_006254257.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
