Assay Details
Gene Symbol: Phyh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053674.1 | NP_446126.1 | 5 - 6 | 504 | 60 | |
GenBank mRNA | AF121345.1 | - | 5 - 6 | 504 | 60 | |
BC086573.1 | - | 5 - 6 | 499 | 60 | ||
FQ216943.1 | - | 3 - 4 | 336 | 60 | ||
FQ224924.1 | - | 3 - 4 | 336 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
