Assay Details
Gene Symbol: Sec61a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170343.1 | NP_001163814.1 | 8 - 9 | 850 | 67 | |
XM_006254257.2 | XP_006254319.1 | 6 - 7 | 686 | 67 | ||
GenBank mRNA | CD373126.1 | - | 4 - 5 | 531 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
