Assay Details
Gene Symbol: Sec61a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170343.1 | NP_001163814.1 | 11 - 12 | 1320 | 81 | |
XM_006254257.2 | XP_006254319.1 | 9 - 10 | 1156 | 81 | ||
GenBank mRNA | CO567453.1 | - | 496 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
