Assay Details
Gene Symbol: Sec61a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170343.1 | NP_001163814.1 | 5 - 6 | 420 | 71 | |
GenBank mRNA | CD373126.1 | - | 1 - 2 | 101 | 71 | |
CK470805.1 | - | 5 - 6 | 443 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
