Assay Details
Gene Symbol: Prpf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138523.2 | NP_612532.1 | 4 - 5 | 467 | 88 | |
GenBank mRNA | AF244920.1 | - | 4 - 5 | 462 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
