Assay Details
Gene Symbol: Prpf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138523.2 | NP_612532.1 | 9 - 10 | 1048 | 116 | |
GenBank mRNA | AF244920.1 | - | 9 - 10 | 1043 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P491929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722090 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370582 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102065 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169045 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P491929 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P491929 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P491929 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P491929 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702134670 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11827 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 55378 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 85815 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 702134670 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10796378 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796373 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796374 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796367 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796368 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796372 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796376 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796375 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796382 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796377 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796370 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796380 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796379 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796371 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796369 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10796366 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10796381 | - | - |
Rat Genome 230 2.0 Array | 1388005_PM_at | - | - |
Rat Genome 230 2.0 Array | 1399021_PM_at | - | - |
Rat Genome 230 2.0 Array | 1388005_at | - | - |
Rat Genome 230 2.0 Array | 1399021_at | - | - |
Affymetrix HT Rat Focus Array | 1388005_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388005_at | - | - |
Affymetrix HT Rat Focus Array | 1399021_at | - | - |
Affymetrix HT Rat Focus Array | 1399021_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1700006961.rn.1 | TC1700001008.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700006962.rn.1 | TC1700001008.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700006954.rn.1 | TC1700001008.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700006958.rn.1 | TC1700001008.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700006960.rn.1 | TC1700001008.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700006956.rn.1 | TC1700001008.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700006957.rn.1 | TC1700001008.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700006959.rn.1 | TC1700001008.rn.1 | RTA TC |
Rat Expression Set 230 | 1388005_PM_at | - | - |
Rat Expression Set 230 | 1388005_at | - | - |
Rat Expression Set 230 | 1399021_PM_at | - | - |
Rat Expression Set 230 | 1399021_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796381 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10796366 | - | - |
Rat Genome U34 Set | rc_AI029421_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21358772 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353121 | - | - |
RatRef-12 Expression BeadChip | ILMN_50750 | - | - |
