Assay Details
Gene Symbol: Sephs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104630.1 | NP_001098100.1 | 5 - 6 | 924 | 49 | |
XM_006254266.2 | XP_006254328.1 | 5 - 6 | 2551 | 49 | ||
GenBank mRNA | AY724491.1 | - | 5 - 6 | 924 | 49 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
