Assay Details
Gene Symbol: Etl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006254331.2 | XP_006254393.1 | 10 - 11 | 3194 | 72 | |
XM_006254332.2 | XP_006254394.1 | 10 - 11 | 2380 | 72 | ||
XM_008771904.1 | XP_008770126.1 | 13 - 14 | 3131 | 72 | ||
XM_008771906.1 | XP_008770128.1 | 12 - 13 | 3026 | 72 | ||
XM_008771908.1 | XP_008770130.1 | 13 - 14 | 3131 | 72 | ||
XM_008771909.1 | XP_008770131.1 | 13 - 14 | 2753 | 72 | ||
XM_008771910.1 | XP_008770132.1 | 13 - 14 | 3133 | 72 | ||
XM_008771911.1 | XP_008770133.1 | 12 - 13 | 3027 | 72 | ||
XM_008771912.1 | XP_008770134.1 | 13 - 14 | 3133 | 72 | ||
XM_008771913.1 | XP_008770135.1 | 12 - 13 | 3026 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
