Assay Details
Gene Symbol: Commd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198732.2 | NP_942027.1 | 3 - 4 | 351 | 119 | |
GenBank mRNA | BC060591.1 | - | 3 - 4 | 354 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
