Assay Details
Gene Symbol: Aga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031641.1 | NP_001026811.1 | 2 - 3 | 285 | 85 | |
XM_006253111.2 | XP_006253173.1 | 3 - 4 | 410 | 85 | ||
GenBank mRNA | BC098718.1 | - | 2 - 3 | 285 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
