Assay Details
Gene Symbol: Aga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031641.1 | NP_001026811.1 | 4 - 5 | 511 | 87 | |
XM_006253111.2 | XP_006253173.1 | 5 - 6 | 636 | 87 | ||
GenBank mRNA | BC098718.1 | - | 4 - 5 | 511 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
