Assay Details
Gene Symbol: Lrp2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017443.1 | NP_001017443.1 | 2 - 3 | 285 | 81 | |
XM_006253141.2 | XP_006253203.1 | 2 - 3 | 201 | 81 | ||
XM_006253143.2 | XP_006253205.1 | 3 - 4 | 1282 | 81 | ||
XM_008771257.1 | XP_008769479.1 | 2 - 3 | 201 | 81 | ||
GenBank mRNA | BC092575.1 | - | 2 - 3 | 285 | 7114719 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
