Assay Details
Gene Symbol: Lrp2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017443.1 | NP_001017443.1 | 4 - 5 | 516 | 134 | |
XM_006253141.2 | XP_006253203.1 | 4 - 5 | 432 | 134 | ||
XM_006253143.2 | XP_006253205.1 | 5 - 6 | 1513 | 134 | ||
GenBank mRNA | BC092575.1 | - | 4 - 5 | 516 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
