Assay Details
Gene Symbol: Enpp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107311.1 | NP_001100781.1 | 4 - 5 | 738 | 98 | |
XM_006253156.2 | XP_006253218.1 | 4 - 5 | 756 | 98 | ||
XM_008771259.1 | XP_008769481.1 | 4 - 5 | 756 | 98 | ||
GenBank mRNA | BC158772.1 | - | 4 - 5 | 718 | 98 |
Gene Symbol: LOC100911549 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767456.1 | XP_008765678.1 | 3 - 4 | 495 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
