Assay Details
Gene Symbol: Casp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012922.2 | NP_037054.1 | 2 - 3 | 112 | 125 | |
XM_006253130.2 | XP_006253192.1 | 2 - 3 | 156 | 125 | ||
GenBank mRNA | BC081854.1 | - | 2 - 3 | 112 | 7104685 | 125 |
U49930.1 | - | 1 - 2 | 82 | 125 | ||
U84410.1 | - | 1 - 2 | 86 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
