Assay Details
Gene Symbol: Vegfc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053653.1 | NP_446105.1 | 5 - 6 | 883 | 121 | |
GenBank mRNA | AY032729.1 | - | 5 - 6 | 883 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
