Assay Details
Gene Symbol: Vegfc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053653.1 | NP_446105.1 | 6 - 7 | 1219 | 88 | |
GenBank mRNA | AY032729.1 | - | 6 - 7 | 1219 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
positive regulation of cell-matrix adhesion
positive regulation of neuroblast proliferation
positive regulation of cell proliferation
negative regulation of cell proliferation
organ morphogenesis
morphogenesis of embryonic epithelium
regulation of vascular endothelial growth factor receptor signaling pathway
positive regulation of protein autophosphorylation
response to drug
positive regulation of blood vessel endothelial cell migration
regulation of angiogenesis
positive regulation of angiogenesis
negative regulation of blood pressure
vascular endothelial growth factor receptor signaling pathway
positive regulation of epithelial cell proliferation
positive regulation of protein secretion
positive regulation of peptidyl-tyrosine phosphorylation
positive chemotaxis
induction of positive chemotaxis
positive regulation of cell division
positive regulation of mast cell chemotaxis
positive regulation of lymphangiogenesis
growth factor activity
chemoattractant activity
vascular endothelial growth factor receptor 3 binding
angiogenesis
positive regulation of cell-matrix adhesion
positive regulation of neuroblast proliferation
positive regulation of cell proliferation
negative regulation of cell proliferation
organ morphogenesis
morphogenesis of embryonic epithelium
regulation of vascular endothelial growth factor receptor signaling pathway
positive regulation of protein autophosphorylation
response to drug
positive regulation of blood vessel endothelial cell migration
regulation of angiogenesis
positive regulation of angiogenesis
negative regulation of blood pressure
vascular endothelial growth factor receptor signaling pathway
positive regulation of epithelial cell proliferation
positive regulation of protein secretion
positive regulation of peptidyl-tyrosine phosphorylation
positive chemotaxis
induction of positive chemotaxis
positive regulation of cell division
positive regulation of mast cell chemotaxis
positive regulation of lymphangiogenesis
growth factor activity
chemoattractant activity
vascular endothelial growth factor receptor 3 binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P14264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251545 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442183 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485936 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922344 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120282 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256872 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907817 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135236 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82014 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93408 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14264 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14264 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P14264 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P14264 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 54797 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 17789 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 30000 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 70555 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62618 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 63877 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53778 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 5315 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10791572 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791569 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791570 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791566 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791567 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791571 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791568 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10791565 | - | - |
Rat Genome 230 2.0 Array | 1368463_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368463_at | - | - |
Affymetrix HT Rat Focus Array | 1368463_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368463_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1600011976.rn.1 | TC1600001585.rn.1 | RTA TC |
Rat Expression Set 230 | 1368463_at | - | - |
Rat Expression Set 230 | 1368463_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10791565 | - | - |
Rat Genome U34 Set | rc_AA899702_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21222905 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349830 | - | - |
RatRef-12 Expression BeadChip | ILMN_68401 | - | - |
