Assay Details
Gene Symbol: Ufsp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014142.1 | NP_001014164.1 | 10 - 11 | 1236 | 68 | |
XM_006253159.2 | XP_006253221.1 | 10 - 11 | 1226 | 68 | ||
XM_008771265.1 | XP_008769487.1 | 10 - 11 | 1387 | 68 | ||
GenBank mRNA | BC083771.1 | - | 10 - 11 | 1236 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
