Assay Details
Gene Symbol: Ufsp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014142.1 | NP_001014164.1 | 2 - 3 | 119 | 95 | |
XM_006253159.2 | XP_006253221.1 | 2 - 3 | 134 | 95 | ||
XM_008771265.1 | XP_008769487.1 | 2 - 3 | 270 | 95 | ||
GenBank mRNA | BC083771.1 | - | 2 - 3 | 119 | 95 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_006253159.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
