Assay Details
Gene Symbol: Ufsp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014142.1 | NP_001014164.1 | 9 - 10 | 1154 | 70 | |
XM_006253159.2 | XP_006253221.1 | 9 - 10 | 1144 | 70 | ||
XM_008771265.1 | XP_008769487.1 | 9 - 10 | 1305 | 70 | ||
GenBank mRNA | BC083771.1 | - | 9 - 10 | 1154 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
