Assay Details
Gene Symbol: Pdlim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053650.1 | NP_446102.1 | 2 - 3 | 325 | 114 | |
XM_006253119.2 | XP_006253181.1 | 2 - 3 | 360 | 117 | ||
XM_006253120.2 | XP_006253182.1 | 2 - 3 | 358 | 117 | ||
GenBank mRNA | AF002281.1 | - | 2 - 3 | 343 | 114 | |
BC081703.1 | - | 2 - 3 | 317 | 117 | ||
FQ215471.1 | - | 2 - 3 | 340 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
